Assay Details
| Gene Symbol: LOC4331445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775493.2 | XP_015630979.1 | 4 - 5 | 579 | 106 | |
| GenBank mRNA | AB003328.1 | - | 4 - 5 | 487 | 106 | |
| AK067641.1 | - | 4 - 5 | 507 | 106 | ||
| AK104921.1 | - | 3 - 4 | 399 | 106 | ||
| AY332476.1 | - | 3 - 4 | 394 | 106 | ||
| AY551914.1 | - | 2 - 3 | 197 | 106 | ||
More Information
Set Membership: |
|

LOC4331445
Genomic Map