Assay Details
Gene Symbol: LOC4333337 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015774816.2 | XP_015630302.1 | 18 - 19 | 3883 | 132 | |
XM_015774817.2 | XP_015630303.1 | 17 - 18 | 3757 | 132 | ||
GenBank mRNA | AK066794.1 | - | 4 - 5 | 1008 | 132 | |
AK072334.1 | - | 18 - 19 | 3889 | 132 |
Gene Symbol: LOC4346696 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015756031.2 | XP_015611517.1 | 17 - 18 | 3774 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() |
