Assay Details
Gene Symbol: LOC4333337 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015774816.2 | XP_015630302.1 | 9 - 10 | 1663 | 105 | |
XM_015774817.2 | XP_015630303.1 | 8 - 9 | 1537 | 105 | ||
GenBank mRNA | AK066278.1 | - | 9 - 10 | 1543 | 105 | |
AK072334.1 | - | 9 - 10 | 1669 | 105 |
Gene Symbol: LOC4346696 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015756031.2 | XP_015611517.1 | 9 - 10 | 1641 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
