Assay Details
Gene Symbol: LOC4333378 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775737.2 | XP_015631223.1 | 1 - 2 | 176 | 108 | |
GenBank mRNA | AK073607.1 | - | 1 - 2 | 141 | 108 | |
AK099029.1 | - | 1 - 2 | 140 | 108 | ||
AK101999.1 | - | 1 - 2 | 95 | 108 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015775737.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
