Assay Details
Gene Symbol: LOC4333378 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775737.2 | XP_015631223.1 | 3 - 4 | 1228 | 111 | |
GenBank mRNA | AK073607.1 | - | 3 - 4 | 1193 | 111 | |
AK099029.1 | - | 3 - 4 | 1192 | 111 | ||
AK101999.1 | - | 3 - 4 | 1147 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
