Assay Details
Gene Symbol: LOC4333378 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775737.2 | XP_015631223.1 | 4 - 5 | 1304 | 126 | |
GenBank mRNA | AK073607.1 | - | 4 - 5 | 1269 | 126 | |
AK099029.1 | - | 4 - 5 | 1268 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
