Assay Details
Gene Symbol: LOC4333378 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775737.2 | XP_015631223.1 | 9 - 10 | 1990 | 108 | |
GenBank mRNA | AK073607.1 | - | 9 - 10 | 1955 | 108 | |
AK099029.1 | - | 9 - 10 | 1954 | 108 | ||
AK101999.1 | - | 9 - 10 | 1906 | 108 | ||
AK104029.1 | - | 1 - 2 | 228 | 108 | ||
BT129248.1 | - | 1 - 2 | 56 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
