Assay Details
Gene Symbol: LOC9272009 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015774351.2 | XP_015629837.1 | 4 - 5 | 514 | 88 | |
GenBank mRNA | AK063769.1 | - | 1711 | 88 | ||
AK241140.1 | - | 4 - 5 | 521 | 88 | ||
AK287904.1 | - | 4 - 5 | 471 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
