Assay Details
Gene Symbol: LOC9272009 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015774351.2 | XP_015629837.1 | 5 - 6 | 612 | 69 | |
GenBank mRNA | AK063769.1 | - | 1809 | 69 | ||
AK241140.1 | - | 5 - 6 | 619 | 69 | ||
AK287904.1 | - | 5 - 6 | 569 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
