Assay Details
Gene Symbol: LOC9272009 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015774351.2 | XP_015629837.1 | 8 - 9 | 972 | 60 | |
GenBank mRNA | AK063769.1 | - | 2169 | 60 | ||
AK241140.1 | - | 8 - 9 | 979 | 60 | ||
AK287904.1 | - | 8 - 9 | 929 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
