Assay Details
| Gene Symbol: LOC4334964 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777950.2 | XP_015633436.1 | 4 - 5 | 649 | 76 | |
| XM_015777951.2 | XP_015633437.1 | 5 - 6 | 1005 | 76 | ||
| XM_015777952.2 | XP_015633438.1 | 5 - 6 | 980 | 76 | ||
| GenBank mRNA | AK102071.1 | - | 4 - 5 | 567 | 76 | |
More Information
Set Membership: |
|

LOC4334964
Genomic Map