Assay Details
Gene Symbol: LOC4335856 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015778019.2 | XP_015633505.1 | 1 - 2 | 362 | 87 | |
GenBank mRNA | AK073414.1 | - | 1 - 2 | 296 | 87 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015778019.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
