Assay Details
| Gene Symbol: LOC4335968 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777910.2 | XP_015633396.1 | 7 - 8 | 1234 | 135 | |
| GenBank mRNA | AK073743.1 | - | 7 - 8 | 1239 | 135 | |
| AK103814.1 | - | 7 - 8 | 1237 | 135 | ||
More Information
Set Membership: |
|

LOC4335968
Genomic Map