Assay Details
| Gene Symbol: LOC4336388 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778859.2 | XP_015634345.1 | 6 - 7 | 735 | 80 | |
| GenBank mRNA | AK067841.1 | - | 6 - 7 | 742 | 80 | |
More Information
Set Membership: |
|

LOC4336388
Genomic Map