Assay Details
Gene Symbol: LOC4336481 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780364.2 | XP_015635850.1 | 6 - 7 | 1063 | 58 | |
GenBank mRNA | BT129736.1 | - | 3 - 4 | 209 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
