Assay Details
| Gene Symbol: LOC4336874 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779927.2 | XP_015635413.1 | 3 - 4 | 1055 | 53 | |
| GenBank mRNA | AB004864.1 | - | 3 - 4 | 1033 | 53 | |
| AB007452.1 | - | 3 - 4 | 1026 | 53 | ||
| AK064040.1 | - | 3 - 4 | 1045 | 53 | ||
More Information
Set Membership: |
|

LOC4336874
Genomic Map