Assay Details
Gene Symbol: LOC4336993 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780527.2 | XP_015636013.1 | 1 - 2 | 477 | 142 | |
GenBank mRNA | AK072027.1 | - | 1 - 2 | 435 | 142 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
