Assay Details
Gene Symbol: LOC4337289 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780728.2 | XP_015636214.1 | 13 - 14 | 2851 | 111 | |
GenBank mRNA | AK066532.1 | - | 13 - 14 | 2857 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
