Assay Details
Gene Symbol: LOC4337289 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780728.2 | XP_015636214.1 | 7 - 8 | 1519 | 81 | |
GenBank mRNA | AK066532.1 | - | 7 - 8 | 1525 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
