Assay Details
Gene Symbol: LOC4337637 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015781970.2 | XP_015637456.1 | 1 - 2 | 513 | 82 | |
GenBank mRNA | AK067943.1 | - | 1 - 2 | 291 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
