Assay Details
| Gene Symbol: LOC4337736 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782285.2 | XP_015637771.1 | 1 - 2 | 581 | 114 | |
| GenBank mRNA | AK100498.1 | - | 1 - 2 | 274 | 114 | |
| EU703799.1 | - | 1 - 2 | 237 | 114 | ||
More Information
Set Membership: |
|

LOC4337736
Genomic Map