Assay Details
| Gene Symbol: LOC4337736 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782285.2 | XP_015637771.1 | 2 - 3 | 653 | 74 | |
| GenBank mRNA | AK059282.1 | - | 2 - 3 | 391 | 74 | |
| AK100498.1 | - | 2 - 3 | 346 | 74 | ||
| EU703799.1 | - | 2 - 3 | 309 | 74 | ||
More Information
Set Membership: |
|

LOC4337736
Genomic Map