Assay Details
| Gene Symbol: LOC4338052 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782828.2 | XP_015638314.1 | 2 - 3 | 315 | 109 | |
| XM_015782829.2 | XP_015638315.1 | 2 - 3 | 332 | 109 | ||
| GenBank mRNA | AK287942.1 | - | 2 - 3 | 364 | 109 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015782828.2, XM_015782829.2 transcripts.
Set Membership: |
|

LOC4338052
Genomic Map