Assay Details
| Gene Symbol: LOC4338349 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784812.2 | XP_015640298.1 | 2 - 3 | 239 | 126 | |
| XM_015784813.2 | XP_015640299.1 | 2 - 3 | 92 | 126 | ||
| GenBank mRNA | AK073928.1 | - | 2 - 3 | 244 | 126 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015784812.2, XM_015784813.2 transcripts.
Set Membership: |
|

LOC4338349
Genomic Map