Assay Details
| Gene Symbol: LOC4339083 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782587.2 | XP_015638073.1 | 1 | 858 | 146 | |
| GenBank mRNA | AK287651.1 | - | 1 | 608 | 146 | |
More Information
Set Membership: |
|

LOC4339083
Genomic Map