Assay Details
Gene Symbol: LOC4339494 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015784707.2 | XP_015640193.2 | 12 - 13 | 1508 | 91 | |
GenBank mRNA | AK061952.1 | - | 8 - 9 | 814 | 91 | |
AK066240.1 | - | 11 - 12 | 1505 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
