Assay Details
Gene Symbol: LOC4339639 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015784085.2 | XP_015639571.1 | 10 - 11 | 1251 | 78 | |
GenBank mRNA | AK066453.1 | - | 9 - 10 | 1773 | 78 | |
AK067742.1 | - | 10 - 11 | 1142 | 78 | ||
AK098906.1 | - | 10 - 11 | 1139 | 78 | ||
AK098911.1 | - | 10 - 11 | 1144 | 78 | ||
AK100749.1 | - | 5 - 6 | 350 | 78 | ||
AK103421.1 | - | 10 - 11 | 1142 | 78 | ||
AK104838.1 | - | 10 - 11 | 1141 | 78 | ||
AK243002.1 | - | 77 | ||||
AK289096.1 | - | 3963 | 78 | |||
EU267973.1 | - | 10 - 11 | 1129 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
