Assay Details
Gene Symbol: LOC4339639 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015784085.2 | XP_015639571.1 | 11 - 12 | 1408 | 85 | |
GenBank mRNA | AK066453.1 | - | 10 - 11 | 1930 | 85 | |
AK067742.1 | - | 11 - 12 | 1299 | 85 | ||
AK098906.1 | - | 11 - 12 | 1296 | 85 | ||
AK098911.1 | - | 11 - 12 | 1301 | 85 | ||
AK100749.1 | - | 6 - 7 | 507 | 85 | ||
AK103421.1 | - | 11 - 12 | 1299 | 85 | ||
AK104838.1 | - | 11 - 12 | 1298 | 85 | ||
AK243002.1 | - | 4119 | 85 | |||
AK289096.1 | - | 4120 | 85 | |||
EU267973.1 | - | 11 - 12 | 1286 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
