Assay Details
Gene Symbol: LOC4339639 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015784085.2 | XP_015639571.1 | 1 - 2 | 166 | 88 | |
GenBank mRNA | AK067742.1 | - | 1 - 2 | 57 | 88 | |
AK098906.1 | - | 1 - 2 | 54 | 88 | ||
AK098911.1 | - | 1 - 2 | 59 | 88 | ||
AK103421.1 | - | 1 - 2 | 57 | 88 | ||
AK104838.1 | - | 1 - 2 | 56 | 88 | ||
AK243002.1 | - | 2878 | 88 | |||
AK289096.1 | - | 2878 | 88 | |||
EU267973.1 | - | 1 - 2 | 44 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015784085.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
