Assay Details
Gene Symbol: LOC4339639 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015784085.2 | XP_015639571.1 | 3 - 4 | 590 | 72 | |
GenBank mRNA | AK066453.1 | - | 2 - 3 | 1112 | 72 | |
AK067742.1 | - | 3 - 4 | 481 | 72 | ||
AK098906.1 | - | 3 - 4 | 478 | 72 | ||
AK098911.1 | - | 3 - 4 | 483 | 72 | ||
AK103421.1 | - | 3 - 4 | 481 | 72 | ||
AK104838.1 | - | 3 - 4 | 480 | 72 | ||
AK243002.1 | - | 3302 | 72 | |||
AK289096.1 | - | 3302 | 72 | |||
EU267973.1 | - | 3 - 4 | 468 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
