Assay Details
Gene Symbol: LOC4339639 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015784085.2 | XP_015639571.1 | 5 - 6 | 803 | 67 | |
GenBank mRNA | AK066453.1 | - | 4 - 5 | 1325 | 67 | |
AK067742.1 | - | 5 - 6 | 694 | 67 | ||
AK098906.1 | - | 5 - 6 | 691 | 67 | ||
AK098911.1 | - | 5 - 6 | 696 | 67 | ||
AK103421.1 | - | 5 - 6 | 694 | 67 | ||
AK104838.1 | - | 5 - 6 | 693 | 67 | ||
AK243002.1 | - | 3515 | 67 | |||
AK289096.1 | - | 3515 | 67 | |||
EU267973.1 | - | 5 - 6 | 681 | 67 |
Gene Symbol: LOC4339221 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | L12252.1 | - | 715 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
