Assay Details
Gene Symbol: LOC4339639 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015784085.2 | XP_015639571.1 | 8 - 9 | 1074 | 67 | |
GenBank mRNA | AK066453.1 | - | 7 - 8 | 1596 | 67 | |
AK067742.1 | - | 8 - 9 | 965 | 67 | ||
AK098906.1 | - | 8 - 9 | 962 | 67 | ||
AK098911.1 | - | 8 - 9 | 967 | 67 | ||
AK100749.1 | - | 3 - 4 | 173 | 67 | ||
AK103421.1 | - | 8 - 9 | 965 | 67 | ||
AK104838.1 | - | 8 - 9 | 964 | 67 | ||
AK243002.1 | - | 3786 | 67 | |||
AK289096.1 | - | 3786 | 67 | |||
EU267973.1 | - | 8 - 9 | 952 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
