Assay Details
Gene Symbol: LOC4339861 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015788058.2 | XP_015643544.1 | 5 - 6 | 2939 | 86 | |
XM_015788059.2 | XP_015643545.1 | 6 - 7 | 2811 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
