Assay Details
| Gene Symbol: LOC4339861 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788058.2 | XP_015643544.1 | 7 - 8 | 3202 | 114 | |
| XM_015788059.2 | XP_015643545.1 | 8 - 9 | 3074 | 114 | ||
| GenBank mRNA | AK106556.1 | - | 3 - 4 | 330 | 114 | |
More Information
Set Membership: |
|

LOC4339861
Genomic Map