Assay Details
Gene Symbol: LOC4339861 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015788058.2 | XP_015643544.1 | 11 - 12 | 3871 | 98 | |
XM_015788059.2 | XP_015643545.1 | 12 - 13 | 3743 | 98 | ||
GenBank mRNA | AK106556.1 | - | 7 - 8 | 999 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
