Assay Details
Gene Symbol: LOC4339861 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015788058.2 | XP_015643544.1 | 12 - 13 | 4162 | 85 | |
XM_015788059.2 | XP_015643545.1 | 13 - 14 | 4034 | 85 | ||
GenBank mRNA | AK106556.1 | - | 8 - 9 | 1290 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
