Assay Details
Gene Symbol: LOC4339866 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015788730.2 | XP_015644216.1 | 1 - 2 | 245 | 109 | |
GenBank mRNA | AK111784.1 | - | 1 - 2 | 237 | 109 | |
AK112060.1 | - | 1 - 2 | 228 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
