Assay Details
Gene Symbol: LOC4339866 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015788730.2 | XP_015644216.1 | 2 - 3 | 345 | 63 | |
GenBank mRNA | AK111784.1 | - | 2 - 3 | 337 | 63 | |
AK112060.1 | - | 2 - 3 | 328 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
