Assay Details
Gene Symbol: LOC4339866 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015788730.2 | XP_015644216.1 | 5 - 6 | 572 | 96 | |
GenBank mRNA | AK111784.1 | - | 5 - 6 | 564 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
