Assay Details
Gene Symbol: LOC4339866 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015788730.2 | XP_015644216.1 | 6 - 7 | 677 | 65 | |
GenBank mRNA | AK111784.1 | - | 6 - 7 | 669 | 65 | |
BT129970.1 | - | 1 - 2 | 100 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
