Assay Details
Gene Symbol: LOC4340218 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015786410.2 | XP_015641896.1 | 4 - 5 | 656 | 102 | |
GenBank mRNA | AK064184.1 | - | 4 - 5 | 563 | 102 | |
U78890.1 | - | 4 - 5 | 550 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
