Assay Details
Gene Symbol: LOC4340218 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015786410.2 | XP_015641896.1 | 7 - 8 | 870 | 85 | |
GenBank mRNA | AK064184.1 | - | 7 - 8 | 777 | 85 | |
U78890.1 | - | 7 - 8 | 764 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
