Assay Details
| Gene Symbol: LOC4340448 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015786014.2 | XP_015641500.1 | 2 - 3 | 906 | 106 | |
| GenBank mRNA | AK062542.1 | - | 1 - 2 | 64 | 106 | |
| AY603381.1 | - | 2 - 3 | 823 | 106 | ||
More Information
Set Membership: |
|

LOC4340448
Genomic Map