Assay Details
Gene Symbol: LOC4340448 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015786014.2 | XP_015641500.1 | 4 - 5 | 1093 | 71 | |
GenBank mRNA | AK062542.1 | - | 3 - 4 | 251 | 71 | |
AY603381.1 | - | 4 - 5 | 1010 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
