Assay Details
Gene Symbol: LOC4341067 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015786516.2 | XP_015642002.1 | 3 - 4 | 431 | 109 | |
XM_015786517.2 | XP_015642003.1 | 3 - 4 | 381 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
