Assay Details
| Gene Symbol: LOC4326459 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766332.1 | XP_015621818.1 | 1 - 2 | 350 | 68 | |
| GenBank mRNA | AK243573.1 | - | 1 - 2 | 314 | 68 | |
| AY972084.1 | - | 1 - 2 | 193 | 68 | ||
| EU152208.1 | - | 1 - 2 | 194 | 68 | ||
More Information
Set Membership: |
|

LOC4326459
Genomic Map