Assay Details
Gene Symbol: LOC4326732 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766191.2 | XP_015621677.1 | 9 - 10 | 1088 | 65 | |
XM_015766192.2 | XP_015621678.1 | 9 - 10 | 1087 | 65 | ||
GenBank mRNA | AK064461.1 | - | 4 - 5 | 962 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
