Assay Details
Gene Symbol: LOC4327706 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771264.2 | XP_015626750.1 | 1 - 2 | 151 | 74 | |
GenBank mRNA | AK058837.1 | - | 1 - 2 | 105 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015771264.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
