Assay Details
| Gene Symbol: LOC4325116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766514.2 | XP_015622000.1 | 4 - 5 | 882 | 85 | |
| GenBank mRNA | AK243649.1 | - | 4 - 5 | 841 | 85 | |
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length between 71 and 85  3' Most  Detect Genomic DNA  Oryza sativa  Probe spans exons | 
Panther Classification:
 Back To Top
 Back To Top
 LOC4325116
LOC4325116 Genomic Map
Genomic Map Back To Top
 Back To Top