Assay Details
| Gene Symbol: LOC4325731 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766004.1 | XP_015621490.1 | 19 - 20 | 2670 | 80 | |
| GenBank mRNA | AK105265.1 | - | 19 - 20 | 2670 | 80 | |
More Information
Set Membership: |
|

LOC4325731
Genomic Map